Product Details

SNP ID
rs117398920
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:119178986 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTCAGTTAAACCCAAAAGCTAAT[C/T]CATTAAGTTGTACTATTTTCTTCAG
Phenotype
MIM: 604344
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAN1A1 PubMed Links

Gene Details

Gene
MAN1A1
Gene Name
mannosidase alpha class 1A member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005907.3 3098 UTR 3 NP_005898.2
XM_005266986.4 3098 UTR 3 XP_005267043.1
XM_011535833.2 3098 UTR 3 XP_011534135.1
XM_011535834.2 3098 Intron XP_011534136.1

View Full Product Details