Product Details
- SNP ID
-
rs117943173
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:122447004 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTGACTATTGTTTGAAGTACGGAT[A/G]CTGTATGAAAGAGAGTTTAGGAGGA
- Phenotype
-
MIM: 614548
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SERINC1
PubMed Links
Gene Details
- Gene
- SERINC1
- Gene Name
- serine incorporator 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_020755.3 |
1140 |
Silent Mutation |
AGC,AGT |
S332S |
NP_065806.1 |
View Full Product Details