Product Details

SNP ID
rs116204422
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:127442333 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAACAGCATTTTCCTAACAGGTCC[A/G]TCATTTTATGACCCAGGGTTCCTCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KIAA0408 PubMed Links

Gene Details

Gene
KIAA0408
Gene Name
KIAA0408
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014702.4 7891 UTR 3 NP_055517.3
Gene
SOGA3
Gene Name
SOGA family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012279.2 7891 UTR 3 NP_001012279.1

View Full Product Details