Product Details

SNP ID
rs114125924
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:130370423 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTGGGATGAGGAGGAGGAGGCTG[A/G]TAGGGCACTATGATAGGCAGTTCTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC105378159 PubMed Links

Gene Details

Gene
LOC105378159
Gene Name
translation initiation factor IF-2-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017011599.1 Intron XP_016867088.1
Gene
SAMD3
Gene Name
sterile alpha motif domain containing 3
There are no transcripts associated with this gene.

Gene
TMEM200A
Gene Name
transmembrane protein 200A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258276.1 Intron NP_001245205.1
NM_001258277.1 Intron NP_001245206.1
NM_001258278.1 Intron NP_001245207.1
NM_052913.2 Intron NP_443145.1
XM_005266816.2 Intron XP_005266873.1
XM_011535419.2 Intron XP_011533721.1
XM_017010230.1 Intron XP_016865719.1

View Full Product Details