Product Details

SNP ID
rs117859986
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:131644888 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGAGAGGAAAGCCAGGTAACTATT[A/G]TGTTCTAGAAACCAAGTGCAGAAAG
Phenotype
MIM: 602182
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ENPP3 PubMed Links

Gene Details

Gene
ENPP3
Gene Name
ectonucleotide pyrophosphatase/phosphodiesterase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005021.4 Intron NP_005012.2
XM_011535897.1 Intron XP_011534199.1
XM_017010932.1 Intron XP_016866421.1

View Full Product Details