Product Details

SNP ID
rs113891596
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:133988596 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCACTGCACTGAACAACACATAT[A/T]ATTTTTATCTTTTTTCTTCTCAATG
Phenotype
MIM: 610372 MIM: 605521
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
SLC2A12 PubMed Links

Gene Details

Gene
SLC2A12
Gene Name
solute carrier family 2 member 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145176.2 4584 UTR 3 NP_660159.1
XM_006715349.3 4584 Intron XP_006715412.1
XM_017010311.1 4584 Intron XP_016865800.1
Gene
TBPL1
Gene Name
TATA-box binding protein like 1
There are no transcripts associated with this gene.

View Full Product Details