Product Details

SNP ID
rs117758534
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:99273963 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTAATAGGGTGAATAAACATAAAC[A/G]TTTGGAGACCACTGCCTAAATAGGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAXC PubMed Links

Gene Details

Gene
FAXC
Gene Name
failed axon connections homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032511.2 8714 UTR 3 NP_115900.1
XM_006715582.2 8714 UTR 3 XP_006715645.1
XM_011536188.2 8714 UTR 3 XP_011534490.1
XM_011536189.2 8714 Intron XP_011534491.1

View Full Product Details