Product Details

SNP ID
rs116167350
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:139161514 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTTGTCTGTGTTCCATGTGTGCA[C/T]TTCTGTTAGACCATTAGAGCTCTTA
Phenotype
MIM: 607977 MIM: 611438
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HECA PubMed Links

Gene Details

Gene
HECA
Gene Name
hdc homolog, cell cycle regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016217.2 Intron NP_057301.1
Gene
TXLNB
Gene Name
taxilin beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153235.3 Intron NP_694967.3
XM_005266836.1 Intron XP_005266893.1
XM_011535505.2 Intron XP_011533807.1
XM_011535506.2 Intron XP_011533808.1
XM_017010322.1 Intron XP_016865811.1

View Full Product Details