Product Details

SNP ID
rs111329046
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:145864864 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAGTGAGCAAATGTATTAATGCCA[A/C]TATCATTTTTTGCTAATGTTCTTCT
Phenotype
MIM: 609101 MIM: 608048
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FBXO30 PubMed Links

Gene Details

Gene
FBXO30
Gene Name
F-box protein 30
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032145.4 Intron NP_115521.3
XM_005267159.3 Intron XP_005267216.1
XM_011536177.2 Intron XP_011534479.1
XM_017011353.1 Intron XP_016866842.1
Gene
LOC100507557
Gene Name
uncharacterized LOC100507557
There are no transcripts associated with this gene.

Gene
SHPRH
Gene Name
SNF2 histone linker PHD RING helicase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042683.2 Intron NP_001036148.2
NM_173082.3 Intron NP_775105.1
XM_006715439.3 Intron XP_006715502.1
XM_006715441.3 Intron XP_006715504.1
XM_006715443.3 Intron XP_006715506.1
XM_011535719.2 Intron XP_011534021.1
XM_011535722.2 Intron XP_011534024.1
XM_017010691.1 Intron XP_016866180.1
XM_017010692.1 Intron XP_016866181.1
XM_017010693.1 Intron XP_016866182.1
XM_017010694.1 Intron XP_016866183.1
XM_017010695.1 Intron XP_016866184.1
XM_017010696.1 Intron XP_016866185.1

View Full Product Details