Product Details

SNP ID
rs117731454
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:158654176 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCTCCCTGGTCCCACCAGCCCTC[C/T]CTGCTCCTGTGTGGAGTGACTCTTG
Phenotype
MIM: 601554
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DYNLT1 PubMed Links

Gene Details

Gene
DYNLT1
Gene Name
dynein light chain Tctex-type 1
There are no transcripts associated with this gene.

Gene
SYTL3
Gene Name
synaptotagmin like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009991.3 Intron NP_001009991.2
NM_001242384.1 Intron NP_001229313.1
NM_001242394.1 Intron NP_001229323.1
NM_001242395.1 Intron NP_001229324.1
NM_001318745.1 Intron NP_001305674.1
XM_005267215.4 Intron XP_005267272.1
XM_005267218.4 Intron XP_005267275.1
XM_005267222.4 Intron XP_005267279.1
XM_006715605.3 Intron XP_006715668.1
XM_006715606.3 Intron XP_006715669.1
XM_006715611.3 Intron XP_006715674.1
XM_011536254.2 Intron XP_011534556.1
XM_011536255.2 Intron XP_011534557.1
XM_017011495.1 Intron XP_016866984.1
XM_017011496.1 Intron XP_016866985.1
XM_017011497.1 Intron XP_016866986.1
XM_017011498.1 Intron XP_016866987.1
XM_017011499.1 Intron XP_016866988.1

View Full Product Details