Product Details

SNP ID
rs115834919
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:158843976 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATACACCCCAAACCTAAGTGAAAG[A/G]TTGAGCCTGTAAATTAGGATTCTGA
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
OSTCP1 PubMed Links

Gene Details

Gene
OSTCP1
Gene Name
oligosaccharyltransferase complex subunit pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details