Product Details

SNP ID
rs117287865
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:169436165 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCATTTAATTAATTCATCACAAAT[A/C]TAGAAATCAACCCAAATATCTCTTA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LOC401287 PubMed Links

Gene Details

Gene
LOC401287
Gene Name
uncharacterized LOC401287
There are no transcripts associated with this gene.

Gene
WDR27
Gene Name
WD repeat domain 27
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001202550.1 Intron NP_001189479.1
NM_182552.4 Intron NP_872358.4
XM_011535682.2 Intron XP_011533984.1
XM_011535684.2 Intron XP_011533986.1
XM_011535685.2 Intron XP_011533987.1
XM_011535687.2 Intron XP_011533989.1
XM_011535688.2 Intron XP_011533990.1
XM_011535689.2 Intron XP_011533991.1
XM_011535690.2 Intron XP_011533992.1
XM_011535691.2 Intron XP_011533993.1
XM_011535692.2 Intron XP_011533994.1
XM_011535693.2 Intron XP_011533995.1
XM_011535694.2 Intron XP_011533996.1
XM_011535696.2 Intron XP_011533998.1
XM_011535697.2 Intron XP_011533999.1
XM_017010658.1 Intron XP_016866147.1
XM_017010659.1 Intron XP_016866148.1
XM_017010660.1 Intron XP_016866149.1
XM_017010661.1 Intron XP_016866150.1
XM_017010662.1 Intron XP_016866151.1
XM_017010663.1 Intron XP_016866152.1
XM_017010664.1 Intron XP_016866153.1
XM_017010665.1 Intron XP_016866154.1
XM_017010666.1 Intron XP_016866155.1
XM_017010667.1 Intron XP_016866156.1
XM_017010668.1 Intron XP_016866157.1
XM_017010669.1 Intron XP_016866158.1
XM_017010670.1 Intron XP_016866159.1
XM_017010671.1 Intron XP_016866160.1
XM_017010672.1 Intron XP_016866161.1
XM_017010673.1 Intron XP_016866162.1
XM_017010674.1 Intron XP_016866163.1
XM_017010675.1 Intron XP_016866164.1

View Full Product Details