Product Details

SNP ID
rs113015559
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:169753885 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAGGTATTAATAGGGGACCCTATT[A/G]CCACATGTCTTTCTCCCTCAGTGTA
Phenotype
MIM: 615532 MIM: 186977
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ERMARD PubMed Links

Gene Details

Gene
ERMARD
Gene Name
ER membrane associated RNA degradation
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278531.1 1154 Missense Mutation ACC,GCC T10A NP_001265460.1
NM_001278532.1 1154 Intron NP_001265461.1
NM_001278533.1 1154 Missense Mutation ACC,GCC T10A NP_001265462.1
NM_018341.2 1154 Missense Mutation ACC,GCC T10A NP_060811.1
XM_011535938.2 1154 Missense Mutation ACC,GCC T10A XP_011534240.1
XM_011535939.2 1154 UTR 5 XP_011534241.1
XM_011535940.2 1154 UTR 5 XP_011534242.1
XM_017011027.1 1154 UTR 5 XP_016866516.1
XM_017011028.1 1154 UTR 5 XP_016866517.1
XM_017011029.1 1154 UTR 5 XP_016866518.1
XM_017011030.1 1154 Intron XP_016866519.1
XM_017011031.1 1154 Intron XP_016866520.1
XM_017011032.1 1154 UTR 5 XP_016866521.1
XM_017011033.1 1154 UTR 5 XP_016866522.1
Gene
TCTE3
Gene Name
t-complex-associated-testis-expressed 3
There are no transcripts associated with this gene.

View Full Product Details