Product Details

SNP ID
rs114064242
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:105140666 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAAAAATTAATGGACTTTTTTAAA[A/G]AGCAGTTCAAGTTTACAGATGACTG
Phenotype
MIM: 604577
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BVES PubMed Links

Gene Details

Gene
BVES
Gene Name
blood vessel epicardial substance
There are no transcripts associated with this gene.

Gene
BVES-AS1
Gene Name
BVES antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details