Product Details

SNP ID
rs112392899
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:20033950 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAAATTGTGTAAAACACAGAAAGA[A/T]TTTTATCTACCGAGTATCTGCTGAA
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
OR4K13 PubMed Links

Gene Details

Gene
OR4K13
Gene Name
olfactory receptor family 4 subfamily K member 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004714.1 809 Missense Mutation AAT,ATT N270I NP_001004714.1

View Full Product Details