Product Details
- SNP ID
-
rs114515553
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:20060446 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TATGTAAACCCCTGCACTATAGGAC[A/C]ATCATGAGCCACAAGCTGCTAAAGG
- Phenotype
-
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
OR4L1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs148115082] are located under a probe and SNP(s) [rs2775254] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OR4L1
- Gene Name
- olfactory receptor family 4 subfamily L member 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001004717.1 |
402 |
Silent Mutation |
ACA,ACC |
T134T |
NP_001004717.1 |
View Full Product Details