Product Details
- SNP ID
-
rs111711235
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:20770299 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGTCGAGAATTCAGAGAGTACAAAT[A/G]TGATGTCCTCATGAGAGAAAATGAA
- Phenotype
-
MIM: 611582
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
EDDM3B
PubMed Links
Gene Details
- Gene
- EDDM3B
- Gene Name
- epididymal protein 3B
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_022360.4 |
247 |
Missense Mutation |
TAT,TGT |
Y50C |
NP_071755.1 |
View Full Product Details