Product Details

SNP ID
rs111965614
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:52314795 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGTTCACCGACCTGCTCGGGACC[G/T]GCCTCATCAGCCCAGTGGTACTGGC
Phenotype
MIM: 176804
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PTGER2 PubMed Links

Gene Details

Gene
PTGER2
Gene Name
prostaglandin E receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000956.3 498 Missense Mutation GGC,TGC G83C NP_000947.2

View Full Product Details