Product Details

SNP ID
rs112328505
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:61293309 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAAATGACCATGGCAAGGGCAATA[C/T]GCTAGAGCTTAGAAGCCTGGTGGGC
Phenotype
MIM: 605437
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PRKCH PubMed Links

Gene Details

Gene
PRKCH
Gene Name
protein kinase C eta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006255.4 Intron NP_006246.2
XM_011536954.2 Intron XP_011535256.1
XM_011536955.1 Intron XP_011535257.1
XM_011536956.1 Intron XP_011535258.1
XM_011536957.1 Intron XP_011535259.1
XM_017021458.1 Intron XP_016876947.1
XM_017021459.1 Intron XP_016876948.1

View Full Product Details