Product Details

SNP ID
rs113859084
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:22901501 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAACAGGTATTCAATGCAGGTGT[G/T]GATTCTGTTCTCTTCAACTGGTGGC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
RBM23 PubMed Links

Gene Details

Gene
RBM23
Gene Name
RNA binding motif protein 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077351.1 1908 UTR 3 NP_001070819.1
NM_001077352.1 1908 UTR 3 NP_001070820.1
NM_001308044.1 1908 UTR 3 NP_001294973.1
NM_018107.4 1908 UTR 3 NP_060577.3
XM_011536890.2 1908 UTR 3 XP_011535192.1
XM_011536892.2 1908 UTR 3 XP_011535194.1
XM_011536893.2 1908 UTR 3 XP_011535195.1
XM_011536894.2 1908 UTR 3 XP_011535196.1
XM_011536895.2 1908 UTR 3 XP_011535197.1
XM_011536896.2 1908 Intron XP_011535198.1
XM_011536897.2 1908 UTR 3 XP_011535199.1
XM_011536900.1 1908 UTR 3 XP_011535202.1
XM_011536902.1 1908 UTR 3 XP_011535204.1
XM_011536903.1 1908 UTR 3 XP_011535205.1
XM_011536904.1 1908 UTR 3 XP_011535206.1
XM_011536905.1 1908 UTR 3 XP_011535207.1
XM_011536906.1 1908 UTR 3 XP_011535208.1
XM_017021398.1 1908 UTR 3 XP_016876887.1
XM_017021399.1 1908 UTR 3 XP_016876888.1
XM_017021400.1 1908 UTR 3 XP_016876889.1
XM_017021401.1 1908 UTR 3 XP_016876890.1
XM_017021402.1 1908 UTR 3 XP_016876891.1
XM_017021403.1 1908 UTR 3 XP_016876892.1
XM_017021404.1 1908 UTR 3 XP_016876893.1
XM_017021405.1 1908 UTR 3 XP_016876894.1
XM_017021406.1 1908 UTR 3 XP_016876895.1
XM_017021407.1 1908 UTR 3 XP_016876896.1
XM_017021408.1 1908 UTR 3 XP_016876897.1
XM_017021409.1 1908 UTR 3 XP_016876898.1
XM_017021410.1 1908 UTR 3 XP_016876899.1
XM_017021411.1 1908 UTR 3 XP_016876900.1
XM_017021412.1 1908 Intron XP_016876901.1
XM_017021413.1 1908 Intron XP_016876902.1
XM_017021414.1 1908 UTR 3 XP_016876903.1
XM_017021415.1 1908 UTR 3 XP_016876904.1
XM_017021416.1 1908 UTR 3 XP_016876905.1
XM_017021417.1 1908 UTR 3 XP_016876906.1
XM_017021418.1 1908 UTR 3 XP_016876907.1
XM_017021419.1 1908 UTR 3 XP_016876908.1
XM_017021420.1 1908 UTR 3 XP_016876909.1

View Full Product Details