Product Details

SNP ID
rs115661694
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:69203955 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTCCTGTTTCGCAGATGAGGAAA[C/G]TGAGGCTTAGAGAAGTTTGGCAAAT
Phenotype
MIM: 616940
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
EXD2 PubMed Links

Gene Details

Gene
EXD2
Gene Name
exonuclease 3'-5' domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193360.1 154 UTR 5 NP_001180289.1
NM_001193361.1 154 Intron NP_001180290.1
NM_001193362.1 154 UTR 5 NP_001180291.1
NM_001193363.1 154 Intron NP_001180292.1
NM_018199.3 154 Intron NP_060669.1
XM_005267817.3 154 Intron XP_005267874.1
XM_011536908.2 154 Intron XP_011535210.1
XM_017021421.1 154 UTR 5 XP_016876910.1
Gene
LOC105370550
Gene Name
uncharacterized LOC105370550
There are no transcripts associated with this gene.

View Full Product Details