Product Details

SNP ID
rs112068716
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:69572805 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACGCCGACCCCGCTGGGCAGCTG[C/T]TCCTGGCCGAGGCCCGAGGCGGCCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC177 PubMed Links

Gene Details

Gene
CCDC177
Gene Name
coiled-coil domain containing 177
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271507.1 1188 Missense Mutation AAC,AGC N273S NP_001258436.1

View Full Product Details