Product Details

SNP ID
rs112222661
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:73712801 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGTTCACATTATCTTTATCAATG[A/G]CCCCCTTCTCTACCACCTTCTGTAG
Phenotype
MIM: 610062 MIM: 604010
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNAL1 PubMed Links

Gene Details

Gene
DNAL1
Gene Name
dynein axonemal light chain 1
There are no transcripts associated with this gene.

Gene
ELMSAN1
Gene Name
ELM2 and Myb/SANT domain containing 1
There are no transcripts associated with this gene.

Gene
PNMA1
Gene Name
paraneoplastic Ma antigen 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006029.4 1625 Missense Mutation GCC,GTC A280V NP_006020.4

View Full Product Details