Product Details

SNP ID
rs114859538
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:73888481 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCATAGTAAACACATTAGAAGTG[A/G]GAAGTGACCGTTAATGGGCACAGGG
Phenotype
MIM: 608642
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PTGR2 PubMed Links

Gene Details

Gene
PTGR2
Gene Name
prostaglandin reductase 2
There are no transcripts associated with this gene.

Gene
ZNF410
Gene Name
zinc finger protein 410
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242924.1 Intron NP_001229853.1
NM_001242926.1 Intron NP_001229855.1
NM_001242927.1 Intron NP_001229856.1
NM_001242928.1 Intron NP_001229857.1
NM_021188.2 Intron NP_067011.1

View Full Product Details