Product Details

SNP ID
rs115498311
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:74663790 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCATCCTGTGCACCTCTTCATAG[A/G]AGTCATATGTAGGGAGGCACAGCTG
Phenotype
MIM: 615380
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AREL1 PubMed Links

Gene Details

Gene
AREL1
Gene Name
apoptosis resistant E3 ubiquitin protein ligase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039479.1 2694 Missense Mutation TCC,TTC S801F NP_001034568.1
XM_005268244.3 2694 Missense Mutation TCC,TTC S640F XP_005268301.1
XM_006720344.2 2694 Missense Mutation TCC,TTC S791F XP_006720407.1
XM_011537415.2 2694 Missense Mutation TCC,TTC S801F XP_011535717.1
XM_011537416.2 2694 Missense Mutation TCC,TTC S741F XP_011535718.1
XM_011537417.2 2694 Missense Mutation TCC,TTC S640F XP_011535719.1

View Full Product Details