Product Details

SNP ID
rs117133499
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:24161777 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAGAACCTACCTCTGCTGATGCT[C/T]GGGAATCTAGACAGGGCCTGTCACC
Phenotype
MIM: 147574 MIM: 612487
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IRF9 PubMed Links

Gene Details

Gene
IRF9
Gene Name
interferon regulatory factor 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006084.4 Intron NP_006075.3
Gene
RNF31
Gene Name
ring finger protein 31
There are no transcripts associated with this gene.

View Full Product Details