Product Details

SNP ID
rs113738849
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:77458271 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCAACGTCAACAACTGGCACTGG[A/T]GAGGGCCAGAAAAGCAGGCCGGCCT
Phenotype
MIM: 608466 MIM: 613401
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
AHSA1 PubMed Links

Gene Details

Gene
AHSA1
Gene Name
activator of Hsp90 ATPase activity 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321441.1 Intron NP_001308370.1
NM_012111.2 Intron NP_036243.1
Gene
VIPAS39
Gene Name
VPS33B interacting protein, apical-basolateral polarity regulator, spe-39 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193314.1 Intron NP_001180243.1
NM_001193315.1 Intron NP_001180244.1
NM_001193316.1 Intron NP_001180245.1
NM_001193317.1 Intron NP_001180246.1
NM_022067.3 Intron NP_071350.2
XM_011537066.1 Intron XP_011535368.1
XM_017021579.1 Intron XP_016877068.1
XM_017021580.1 Intron XP_016877069.1
XM_017021581.1 Intron XP_016877070.1

View Full Product Details