Product Details

SNP ID
rs113982416
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:88830860 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTATCTCCTAATTAAAACATTCC[A/G]ACTCTTCCTTCATAGTCCTGCTCCT
Phenotype
MIM: 608132
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TTC8 PubMed Links

Gene Details

Gene
TTC8
Gene Name
tetratricopeptide repeat domain 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288781.1 Intron NP_001275710.1
NM_001288782.1 Intron NP_001275711.1
NM_001288783.1 Intron NP_001275712.1
NM_144596.3 Intron NP_653197.2
NM_198309.3 Intron NP_938051.1
NM_198310.3 Intron NP_938052.1
XM_006720035.1 Intron XP_006720098.1
XM_006720037.2 Intron XP_006720100.1
XM_011536432.1 Intron XP_011534734.1
XM_011536433.1 Intron XP_011534735.1
XM_011536434.1 Intron XP_011534736.1
XM_011536435.2 Intron XP_011534737.1
XM_017020977.1 Intron XP_016876466.1
XM_017020978.1 Intron XP_016876467.1

View Full Product Details