Product Details

SNP ID
rs117000189
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:94022602 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTCCAGTAACTCTTTGCGGAATG[C/T]TGAATGAATGCGTTTTGCCTTGTTG
Phenotype
MIM: 608338
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OTUB2 PubMed Links

Gene Details

Gene
OTUB2
Gene Name
OTU deubiquitinase, ubiquitin aldehyde binding 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_023112.3 Intron NP_075601.1
XM_005268055.3 Intron XP_005268112.1
XM_011537148.2 Intron XP_011535450.1

View Full Product Details