Product Details

SNP ID
rs115831106
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:101065469 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTCTGGGGTACGGGGATGGATGGT[C/T]GACCAGTTGGAAAGTAATTGTTTCT
Phenotype
MIM: 611794 MIM: 614057 MIM: 615036 MIM: 615385 MIM: 616376
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MEG9 PubMed Links

Gene Details

Gene
MEG9
Gene Name
maternally expressed 9 (non-protein coding)
There are no transcripts associated with this gene.

Gene
MIR154
Gene Name
microRNA 154
There are no transcripts associated with this gene.

Gene
MIR323B
Gene Name
microRNA 323b
There are no transcripts associated with this gene.

Gene
MIR369
Gene Name
microRNA 369
There are no transcripts associated with this gene.

Gene
MIR377
Gene Name
microRNA 377
There are no transcripts associated with this gene.

Gene
MIR409
Gene Name
microRNA 409
There are no transcripts associated with this gene.

Gene
MIR410
Gene Name
microRNA 410
There are no transcripts associated with this gene.

Gene
MIR412
Gene Name
microRNA 412
There are no transcripts associated with this gene.

Gene
MIR485
Gene Name
microRNA 485
There are no transcripts associated with this gene.

Gene
MIR496
Gene Name
microRNA 496
There are no transcripts associated with this gene.

Gene
MIR541
Gene Name
microRNA 541
There are no transcripts associated with this gene.

Gene
MIR656
Gene Name
microRNA 656
There are no transcripts associated with this gene.

View Full Product Details