Product Details

SNP ID
rs117545930
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:101770708 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAAGAGCTCATCAAGCAAATGAT[A/G]TGAAATCTAAAATTTTCCAGTTCAA
Phenotype
MIM: 601645
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PPP2R5C PubMed Links

Gene Details

Gene
PPP2R5C
Gene Name
protein phosphatase 2 regulatory subunit B'gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001161725.1 Intron NP_001155197.1
NM_001161726.1 Intron NP_001155198.1
NM_002719.3 Intron NP_002710.2
NM_178586.2 Intron NP_848701.1
NM_178587.2 Intron NP_848702.1
XM_005267819.1 Intron XP_005267876.1
XM_005267820.1 Intron XP_005267877.1
XM_005267822.3 Intron XP_005267879.1
XM_005267823.1 Intron XP_005267880.1
XM_005267824.1 Intron XP_005267881.1
XM_005267826.1 Intron XP_005267883.1
XM_005267827.1 Intron XP_005267884.1
XM_011536914.2 Intron XP_011535216.1
XM_011536915.2 Intron XP_011535217.1
XM_011536916.2 Intron XP_011535218.1
XM_011536918.1 Intron XP_011535220.1
XM_011536919.2 Intron XP_011535221.1
XM_011536920.1 Intron XP_011535222.1
XM_011536921.2 Intron XP_011535223.1
XM_017021422.1 Intron XP_016876911.1
XM_017021423.1 Intron XP_016876912.1
XM_017021424.1 Intron XP_016876913.1

View Full Product Details