Product Details

SNP ID
rs117626295
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:103095734 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAGTCTTTTAAGTGTCTCTTTCT[A/G]ATAGGCTTTGCTAGGGTATAGGAAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EXOC3L4 PubMed Links

Gene Details

Gene
EXOC3L4
Gene Name
exocyst complex component 3 like 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077594.1 Intron NP_001071062.1
XM_011537323.2 Intron XP_011535625.1
XM_011537324.2 Intron XP_011535626.1
XM_011537325.2 Intron XP_011535627.1
XM_011537327.2 Intron XP_011535629.1
XM_011537328.2 Intron XP_011535630.1
XM_011537329.2 Intron XP_011535631.1
XM_011537330.2 Intron XP_011535632.1
XM_011537331.1 Intron XP_011535633.1
XM_011537332.2 Intron XP_011535634.1
XM_011537333.2 Intron XP_011535635.1
XM_011537334.1 Intron XP_011535636.1
Gene
LOC107984640
Gene Name
translation initiation factor IF-2-like
There are no transcripts associated with this gene.

View Full Product Details