Product Details

SNP ID
rs111440604
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:149858370 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTATTGCCATTTTCAACTTCTGG[G/T]TTATTTGTGCTGTTTTCAACATCAG
Phenotype
MIM: 180071 MIM: 608886
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PDE6A PubMed Links

Gene Details

Gene
PDE6A
Gene Name
phosphodiesterase 6A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000440.2 5228 UTR 3 NP_000431.2
XM_011537650.2 5228 Intron XP_011535952.1
XM_011537651.2 5228 Intron XP_011535953.1
XM_011537652.1 5228 Intron XP_011535954.1
XM_011537653.2 5228 Intron XP_011535955.1
XM_011537654.2 5228 Intron XP_011535956.1
XM_017009572.1 5228 Intron XP_016865061.1
Gene
PPARGC1B
Gene Name
PPARG coactivator 1 beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172698.1 5228 Intron NP_001166169.1
NM_001172699.1 5228 Intron NP_001166170.1
NM_133263.3 5228 Intron NP_573570.3
XM_005268372.4 5228 Intron XP_005268429.1
XM_011537553.2 5228 Intron XP_011535855.1
XM_011537554.2 5228 Intron XP_011535856.1
XM_011537555.2 5228 Intron XP_011535857.1
XM_011537556.2 5228 Intron XP_011535858.1
XM_011537557.1 5228 Intron XP_011535859.1

View Full Product Details