Product Details

SNP ID
rs116459403
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:140550889 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAGCGGTGGATGTCATCTGCTGC[A/G]TCCCACCGGTGGCTTGAAAGCTCTG
Phenotype
MIM: 602711 MIM: 603483 MIM: 603819
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANKHD1-EIF4EBP3 PubMed Links

Gene Details

Gene
ANKHD1-EIF4EBP3
Gene Name
ANKHD1-EIF4EBP3 readthrough
There are no transcripts associated with this gene.

Gene
APBB3
Gene Name
amyloid beta precursor protein binding family B member 3
There are no transcripts associated with this gene.

Gene
EIF4EBP3
Gene Name
eukaryotic translation initiation factor 4E binding protein 3
There are no transcripts associated with this gene.

Gene
SRA1
Gene Name
steroid receptor RNA activator 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001035235.3 442 Silent Mutation GAC,GAT D174D NP_001030312.2
NM_001253764.1 442 Silent Mutation GAC,GAT D146D NP_001240693.1

View Full Product Details