Product Details

SNP ID
rs112110449
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:150661331 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTACTGACTGCTGACCGCTGACTG[C/T]CTACAGGGACGCCACGCAACTCTCA
Phenotype
MIM: 610735 MIM: 608155
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MYOZ3 PubMed Links

Gene Details

Gene
MYOZ3
Gene Name
myozenin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001122853.2 491 UTR 5 NP_001116325.1
NM_133371.4 491 Intron NP_588612.2
Gene
SYNPO
Gene Name
synaptopodin
There are no transcripts associated with this gene.

View Full Product Details