Product Details

SNP ID
rs111312047
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:140645680 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACATTCGTCTCTTGGCCAAATGC[G/T]GAAGAGAGAGAGGGAGGTGTCTTTA
Phenotype
MIM: 600549 MIM: 602137
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
IK PubMed Links

Gene Details

Gene
IK
Gene Name
IK cytokine, down-regulator of HLA II
There are no transcripts associated with this gene.

Gene
MIR3655
Gene Name
microRNA 3655
There are no transcripts associated with this gene.

Gene
NDUFA2
Gene Name
NADH:ubiquinone oxidoreductase subunit A2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001185012.1 456 UTR 3 NP_001171941.1
NM_002488.4 456 Intron NP_002479.1
Gene
TMCO6
Gene Name
transmembrane and coiled-coil domains 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300980.1 456 Intron NP_001287909.1
NM_001300982.1 456 Intron NP_001287911.1
NM_018502.4 456 Intron NP_060972.3
XM_005268477.1 456 Intron XP_005268534.1
XM_011537663.2 456 Intron XP_011535965.1
XM_011537665.2 456 Intron XP_011535967.1
XM_011537668.2 456 Intron XP_011535970.1
XM_017009617.1 456 Intron XP_016865106.1
XM_017009618.1 456 Intron XP_016865107.1
XM_017009619.1 456 Intron XP_016865108.1
XM_017009620.1 456 Intron XP_016865109.1

View Full Product Details