Product Details
- SNP ID
-
rs115391126
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:155015252 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCAGGAATTGAAAGAAAATGTAGAG[A/T]TAATTTGTAACCTGCAGCAACTGAT
- Phenotype
-
MIM: 609184
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
KIF4B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs17116709] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- KIF4B
- Gene Name
- kinesin family member 4B
There are no transcripts associated with this gene.
View Full Product Details