Product Details

SNP ID
rs113934136
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:141209022 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGAACTTTTTGGTGATGGAGGAA[C/T]TGCAGAGCGGGAGCTTTGTAGGAAA
Phenotype
MIM: 606337 MIM: 606338 MIM: 606339
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PCDHB11 PubMed Links

Gene Details

Gene
PCDHB11
Gene Name
protocadherin beta 11
There are no transcripts associated with this gene.

Gene
PCDHB12
Gene Name
protocadherin beta 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018932.3 304 Silent Mutation CTG,TTG L39L NP_061755.1
Gene
PCDHB13
Gene Name
protocadherin beta 13
There are no transcripts associated with this gene.

View Full Product Details