Product Details

SNP ID
rs116827726
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:144159278 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTCAATAACACAGTTAAAATTAAT[A/T]GGGAAAGTTTTAATAAGCATTCAAA
Phenotype
MIM: 611483
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
YIPF5 PubMed Links

Gene Details

Gene
YIPF5
Gene Name
Yip1 domain family member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024947.3 2083 UTR 3 NP_001020118.1
NM_001271732.1 2083 UTR 3 NP_001258661.1
NM_030799.8 2083 UTR 3 NP_110426.4

View Full Product Details