Product Details

SNP ID
rs116888459
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:49336156 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCCCGCGTAGAAAGCAATGCGAG[A/G]CACGTAGCCGGCAGCGGGCGCCACC
Phenotype
MIM: 615229 MIM: 603872
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C1QL4 PubMed Links

Gene Details

Gene
C1QL4
Gene Name
complement C1q like 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008223.1 1033 Missense Mutation CCT,TCT P108S NP_001008224.1
XM_011538270.2 1033 Missense Mutation CCT,TCT P108S XP_011536572.1
Gene
TROAP
Gene Name
trophinin associated protein
There are no transcripts associated with this gene.

View Full Product Details