Product Details

SNP ID
rs116898862
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:49836953 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGTAGATGACTTTAATATAGGAA[C/T]CTCCCACCAGGCACCGGACACATGT
Phenotype
MIM: 615104
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BCDIN3D PubMed Links

Gene Details

Gene
BCDIN3D
Gene Name
BCDIN3 domain containing RNA methyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181708.2 2339 UTR 3 NP_859059.1
Gene
BCDIN3D-AS1
Gene Name
BCDIN3D antisense RNA 1
There are no transcripts associated with this gene.

Gene
NCKAP5L
Gene Name
NCK associated protein 5 like
There are no transcripts associated with this gene.

View Full Product Details