Product Details
- SNP ID
-
rs115737603
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:53424443 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGCTTTGGGATCTGGAACCTGACC[A/C]AAGACCGGGCACAGGTGGAAATGCA
- Phenotype
-
MIM: 600956
MIM: 189906
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
AMHR2
PubMed Links
Gene Details
- Gene
- AMHR2
- Gene Name
- anti-Mullerian hormone receptor type 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001164690.1 |
285 |
Missense Mutation |
AAA,CAA |
K69Q |
NP_001158162.1 |
NM_001164691.1 |
285 |
Missense Mutation |
AAA,CAA |
K69Q |
NP_001158163.1 |
NM_020547.2 |
285 |
Missense Mutation |
AAA,CAA |
K69Q |
NP_065434.1 |
XM_011538173.1 |
285 |
Missense Mutation |
AAA,CAA |
K69Q |
XP_011536475.1 |
XM_011538174.1 |
285 |
Missense Mutation |
AAA,CAA |
K69Q |
XP_011536476.1 |
XM_011538176.1 |
285 |
Missense Mutation |
AAA,CAA |
K69Q |
XP_011536478.1 |
XM_011538178.1 |
285 |
Missense Mutation |
AAA,CAA |
K69Q |
XP_011536480.1 |
XM_011538179.1 |
285 |
Missense Mutation |
AAA,CAA |
K69Q |
XP_011536481.1 |
XM_011538180.1 |
285 |
UTR 5 |
|
|
XP_011536482.1 |
XM_011538181.1 |
285 |
UTR 5 |
|
|
XP_011536483.1 |
XM_011538183.2 |
285 |
Missense Mutation |
AAA,CAA |
K69Q |
XP_011536485.1 |
XM_011538184.2 |
285 |
Missense Mutation |
AAA,CAA |
K69Q |
XP_011536486.1 |
XM_011538185.1 |
285 |
Missense Mutation |
AAA,CAA |
K69Q |
XP_011536487.1 |
XM_011538186.2 |
285 |
Intron |
|
|
XP_011536488.1 |
XM_011538187.2 |
285 |
Missense Mutation |
AAA,CAA |
K69Q |
XP_011536489.1 |
XM_017019179.1 |
285 |
Missense Mutation |
AAA,CAA |
K69Q |
XP_016874668.1 |
- Gene
- SP1
- Gene Name
- Sp1 transcription factor
There are no transcripts associated with this gene.
View Full Product Details