Product Details

SNP ID
rs111636907
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:55469328 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGAAGTATTGAGCACAGTTACTC[A/C]TTTGCTTAAAGCAACTCTTTCATTC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OR6C70 PubMed Links

Gene Details

Gene
OR6C70
Gene Name
olfactory receptor family 6 subfamily C member 70
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005499.1 811 Nonsense Mutation GGA,TGA G271* NP_001005499.1

View Full Product Details