Product Details

SNP ID
rs114691351
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:55769116 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACGACTAACAATAAAAGATATCTA[C/T]TTAAGGTAATATATGAGGGGACTGG
Phenotype
MIM: 610049
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SARNP PubMed Links

Gene Details

Gene
SARNP
Gene Name
SAP domain containing ribonucleoprotein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033082.3 Intron NP_149073.1

View Full Product Details