Product Details

SNP ID
rs111947844
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:56003903 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACAAGGTAGCCCCCACCGTGGAG[A/G]CCTCTGACCCTTATGCTGATGATCC
Phenotype
MIM: 606239 MIM: 179514 MIM: 606887
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IKZF4 PubMed Links

Gene Details

Gene
IKZF4
Gene Name
IKAROS family zinc finger 4
There are no transcripts associated with this gene.

Gene
LOC105369781
Gene Name
uncharacterized LOC105369781
There are no transcripts associated with this gene.

Gene
RAB5B
Gene Name
RAB5B, member RAS oncogene family
There are no transcripts associated with this gene.

Gene
SUOX
Gene Name
sulfite oxidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000456.2 1386 Missense Mutation ACC,GCC T172A NP_000447.2
NM_001032386.1 1386 Missense Mutation ACC,GCC T172A NP_001027558.1
NM_001032387.1 1386 Missense Mutation ACC,GCC T172A NP_001027559.1
XM_005269112.1 1386 Missense Mutation ACC,GCC T179A XP_005269169.1
XM_017019905.1 1386 Missense Mutation ACC,GCC T179A XP_016875394.1
XM_017019906.1 1386 Missense Mutation ACC,GCC T179A XP_016875395.1
XM_017019907.1 1386 Missense Mutation ACC,GCC T172A XP_016875396.1
XM_017019908.1 1386 Missense Mutation ACC,GCC T172A XP_016875397.1

View Full Product Details