Product Details

SNP ID
rs114008375
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:56230113 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTTCCTCTGGGTGTCCTTGTCTC[C/T]TGCTATCAGGTAATGCCAACCTCAA
Phenotype
MIM: 612104 MIM: 608730
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANKRD52 PubMed Links

Gene Details

Gene
ANKRD52
Gene Name
ankyrin repeat domain 52
There are no transcripts associated with this gene.

Gene
NABP2
Gene Name
nucleic acid binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024068.3 58 Intron NP_076973.1
XM_005269146.2 58 Intron XP_005269203.1
XM_005269147.2 58 Intron XP_005269204.1
XM_005269148.4 58 Intron XP_005269205.1
XM_005269149.4 58 Intron XP_005269206.1
Gene
RNF41
Gene Name
ring finger protein 41
There are no transcripts associated with this gene.

Gene
SLC39A5
Gene Name
solute carrier family 39 member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135195.1 58 Intron NP_001128667.1
NM_173596.2 58 UTR 5 NP_775867.2
XM_005268803.1 58 UTR 5 XP_005268860.1
XM_011538198.1 58 Intron XP_011536500.1
XM_011538199.1 58 UTR 5 XP_011536501.1
XM_011538200.1 58 UTR 5 XP_011536502.1
XM_011538201.1 58 UTR 5 XP_011536503.1
XM_017019185.1 58 Intron XP_016874674.1
XM_017019186.1 58 UTR 5 XP_016874675.1
XM_017019187.1 58 UTR 5 XP_016874676.1

View Full Product Details