Product Details

SNP ID
rs117683816
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:56244483 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACTGACCCCTCCCTCCAGAGCAG[C/T]AGCCATCCTGGCTCTCCACTTTGCA
Phenotype
MIM: 608730
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANKRD52 PubMed Links

Gene Details

Gene
ANKRD52
Gene Name
ankyrin repeat domain 52
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173595.3 Intron NP_775866.2
XM_011538197.2 Intron XP_011536499.1
XM_017019183.1 Intron XP_016874672.1
XM_017019184.1 Intron XP_016874673.1
Gene
SLC39A5
Gene Name
solute carrier family 39 member 5
There are no transcripts associated with this gene.

View Full Product Details