Product Details

SNP ID
rs112815837
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:57128996 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCCGCCGTTGCTCCTGCTGCTGC[C/T]CCTGCTCTCAGCTCTGGTCGCGGCG
Phenotype
MIM: 107770
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LRP1 PubMed Links

Gene Details

Gene
LRP1
Gene Name
LDL receptor related protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002332.2 498 Missense Mutation CCC,CTC P11L NP_002323.2
XM_017019303.1 498 Missense Mutation CCC,CTC P11L XP_016874792.1

View Full Product Details