Product Details

SNP ID
rs114668530
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:57235998 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATATTAGCAGGTGCTGACAAGTGAG[A/G]CAAGAAGCCTGTCACCATTGTCAGC
Phenotype
MIM: 604637 MIM: 138450 MIM: 615521
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NDUFA4L2 PubMed Links

Gene Details

Gene
NDUFA4L2
Gene Name
NDUFA4, mitochondrial complex associated like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020142.3 Intron NP_064527.1
XM_005269033.2 Intron XP_005269090.1
XM_011538573.2 Intron XP_011536875.1
Gene
NXPH4
Gene Name
neurexophilin 4
There are no transcripts associated with this gene.

Gene
SHMT2
Gene Name
serine hydroxymethyltransferase 2
There are no transcripts associated with this gene.

Gene
STAC3
Gene Name
SH3 and cysteine rich domain 3
There are no transcripts associated with this gene.

View Full Product Details