Product Details

SNP ID
rs112372090
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:57244915 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTTGCAGGGAGGAAGGATTCTTA[C/G]CTTGTCATCAGGTGTCTTCTTCTCA
Phenotype
MIM: 138450 MIM: 615521
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NDUFA4L2 PubMed Links

Gene Details

Gene
NDUFA4L2
Gene Name
NDUFA4, mitochondrial complex associated like 2
There are no transcripts associated with this gene.

Gene
R3HDM2
Gene Name
R3H domain containing 2
There are no transcripts associated with this gene.

Gene
SHMT2
Gene Name
serine hydroxymethyltransferase 2
There are no transcripts associated with this gene.

Gene
STAC3
Gene Name
SH3 and cysteine rich domain 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286256.1 Intron NP_001273185.1
NM_001286257.1 Intron NP_001273186.1
NM_145064.2 Intron NP_659501.1
XM_011538126.2 Intron XP_011536428.1

View Full Product Details